Pentafluorphenylphenylmethansulfonat
Lieferant: Apollo Scientific
|
Achtung
|
PC10318-1G
APOSPC10318-1GEA
198
EUR
APOSPC10318-1G
Pentafluorphenylphenylmethansulfonat
Pentafluorphenylphenylmethansulfonat
Formel:
C₁₃H₇F₅O₃S Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05975135 CAS-Nummer: 864450-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...