1-Chlor-1,2-dibrom-2-fluorpropan 97%

Lieferant: Apollo Scientific

PC1032-5G
APOSPC1032-5GEA 170 EUR
APOSPC1032-5G
1-Chlor-1,2-dibrom-2-fluorpropan 97%
1-Chloro-1,2-dibromo-2-fluoropropane
Formel: C₃H₄Br₂ClF
Lagertemperatur: Raumtemperatur
CAS-Nummer: 885276-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR