2-Bromo-6-fluorophenylhydrazine hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: (2-Bromo-6-fluorophenyl)hydrazine hydrochloride

PC10332-5G
APOSPC10332-5GEA 121 EUR
APOSPC10332-5G
2-Bromo-6-fluorophenylhydrazine hydrochloride 97%
2-Bromo-6-fluorophenylhydrazine hydrochloride
Formel: C₆H₆BrFN₂·ClH
Molecular Weight: 241,489 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368676
CAS-Nummer: 1049729-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR