2,6-Difluor-3-methoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC10334-250MG
APOSPC10334-250MGEA
21.9
EUR
APOSPC10334-250MG
2,6-Difluor-3-methoxyphenylboronsäure
2,6-Difluor-3-methoxyphenylboronsäure
Formel:
C₇H₇BF₂O₃ Schmelzpunkt: 112…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657880 CAS-Nummer: 870779-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...