5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC10340-1G
APOSPC10340-1GEA
74.1
EUR
APOSPC10340-1G
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol 95%
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol
Formel:
C₁₀H₄F₆N₂O Schmelzpunkt: 185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140206 CAS-Nummer: 51420-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...