4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-((5-(Trifluoromethyl)pyridin-2-yl)oxy)aniline

PC10341-1G
APOSPC10341-1GEA 102 EUR
APOSPC10341-1G
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline
Formel: C₁₂H₉F₃N₂O
Molecular Weight: 254,21 g/mol
Schmelzpunkt: 44…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03001213
CAS-Nummer: 71422-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR