2-(Aminooxy)-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide hydrochloride

Lieferant: Apollo Scientific

Synonyme: 2-[(2-{[2-(Ammoniooxy)acetyl]amino}ethyl)amino]-3-chloro-5-(trifluoromethyl)pyridine hydrochloride

PC10342-1G
APOSPC10342-1GEA 783 EUR
APOSPC10342-1G
2-(Aminooxy)-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide hydrochloride
2-(Aminooxy)-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide hydrochloride
Formel: C₁₀H₁₃Cl₂F₃N₄O₂
Molecular Weight: 349,14 g/mol
Schmelzpunkt: 157…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01114568
CAS-Nummer: 324009-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR