4-Hydroxy-3-(trifluormethyl)benzonitril
Lieferant: Apollo Scientific
|
Achtung
|
PC10355-500MG
PC10355-5G
PC10355-25G
APOSPC10355-500MGEA
16.1
EUR
APOSPC10355-500MG
APOSPC10355-5G
APOSPC10355-25G
4-Hydroxy-3-(trifluormethyl)benzonitril
4-Hydroxy-3-(trifluormethyl)benzonitril
Formel:
C₈H₄F₃NO Schmelzpunkt: 171…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797882 CAS-Nummer: 124811-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...