3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC10358-250MG
APOSPC10358-250MGEA
69.6
EUR
APOSPC10358-250MG
3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde 98%
3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde
Formel:
C₁₀H₆F₂N₂O Molecular Weight: 208,16 g/mol Schmelzpunkt: 178…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797905 CAS-Nummer: 875664-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...