Methyl-4-fluor-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
PC10370-1G
APOSPC10370-1GEA
0
EUR
APOSPC10370-1G
Methyl-4-fluor-2-benzo[b]thiophencarboxylat
Methyl-4-fluor-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₇FO₂S Schmelzpunkt: 96…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200964 CAS-Nummer: 220180-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...