5-Methyl-3-(trifluoromethyl)-1H-1,2,4-triazole
Lieferant: Apollo Scientific
|
Achtung
|
PC10373-250MG
APOSPC10373-250MGEA
21.9
EUR
APOSPC10373-250MG
5-Methyl-3-(trifluoromethyl)-1H-1,2,4-triazole
5-Methyl-3-(trifluoromethyl)-1H-1,2,4-triazole
Formel:
C₄H₄F₃N₃ Schmelzpunkt: 143…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03463854 CAS-Nummer: 667873-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...