3,5-Difluor-2-methoxyphenylboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3,5-Difluoro-2-methoxyphenyl)boronic acid, 3,5-Difluoro-2-methoxybenzeneboronic acid
PC10378-1G
PC10378-10G
PC10378-5G
PC10378-250MG
APOSPC10378-1GEA
107
EUR
APOSPC10378-1G
APOSPC10378-10G
APOSPC10378-5G
APOSPC10378-250MG
3,5-Difluor-2-methoxyphenylboronsäure 96%
3,5-Difluor-2-methoxyphenylboronsäure
Formel:
C₇H₇BF₂O₃ Schmelzpunkt: 112…115 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03095353 CAS-Nummer: 737000-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...