5-Fluor-2-methylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

PC10379-100G PC10379-25G
APOSPC10379-100GEA 235 EUR
APOSPC10379-100G APOSPC10379-25G
5-Fluor-2-methylphenylboronsäure
5-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
Schmelzpunkt: 148…149 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03095047
CAS-Nummer: 163517-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR