5-Fluor-2-methylphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid
PC10379-100G
PC10379-25G
APOSPC10379-100GEA
235
EUR
APOSPC10379-100G
APOSPC10379-25G
5-Fluor-2-methylphenylboronsäure
5-Fluor-2-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153.95 g/mol Schmelzpunkt: 148…149 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095047 CAS-Nummer: 163517-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...