4-Chlor-3-(trifluormethylphenyl)isocyanat 97%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-3-(trifluoromethyl)phenylisocyanate, 1-Chloro-4-isocyanato-2-(trifluoromethyl)benzene

PC10386-100G
APOSPC10386-100GEA 58 EUR
APOSPC10386-100G
4-Chlor-3-(trifluormethylphenyl)isocyanat 97%
4-Chlor-3-(trifluormethylphenyl)isocyanat

  • May form small amount of sediment.

Formel: C₈H₃ClF₃NO
Molecular Weight: 221.57 g/mol
Siedepunkt: 86…90 °C/14mmHg
Schmelzpunkt: 34…36 °C
Flammpunkt: 99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013874
CAS-Nummer: 327-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR