4-Fluorbenzoylacetonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4-Fluorphenyl)-3-oxopropionsäurenitril, 3-(4-Fluorphenyl)-3-ketopropionsäurenitril
PC10409-1G
PC10409-5G
PC10409-25G
APOSPC10409-1GEA
21.9
EUR
APOSPC10409-1G
APOSPC10409-5G
APOSPC10409-25G
4-Fluorbenzoylacetonitril
4-Fluorbenzoylacetonitril
Formel:
C₉H₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662062 CAS-Nummer: 4640-67-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...