2-Fluor-5-nitro-4-picolin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluor-4-methyl-5-nitropyridin

PC10419-5G
APOSPC10419-5GEA 23.2 EUR
APOSPC10419-5G
2-Fluor-5-nitro-4-picolin 97%
2-Fluor-5-nitro-4-picolin
Formel: C₆H₅FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03095070
CAS-Nummer: 19346-47-5
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR