N-[(3-Fluorophenyl)carbamothioyl]benzamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzoyl-3-(3-fluorophenyl)thiourea

PC10452-5G
APOSPC10452-5GEA 169 EUR
APOSPC10452-5G
N-[(3-Fluorophenyl)carbamothioyl]benzamide
N-[(3-Fluorophenyl)carbamothioyl]benzamide
Formel: C₁₄H₁₁FN₂OS
Molecular Weight: 274,313 g/mol
Schmelzpunkt: 145…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01023683
CAS-Nummer: 82635-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR