N-{[4-(Trifluoromethyl)phenyl]carbamothioyl}benzamide

Lieferant: Apollo Scientific
Achtung

PC10453-25G PC10453-5G
APOSPC10453-25GEA 759 EUR
APOSPC10453-25G APOSPC10453-5G
N-{[4-(Trifluoromethyl)phenyl]carbamothioyl}benzamide
N-{[4-(Trifluoromethyl)phenyl]carbamothioyl}benzamide
Formel: C₁₅H₁₁F₃N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00588815
CAS-Nummer: 117174-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR