2-Methoxy-5-(trifluormethoxy)benzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC10457-100G
PC10457-25G
PC10457-5G
APOSPC10457-100GEA
0
EUR
APOSPC10457-100G
APOSPC10457-25G
APOSPC10457-5G
2-Methoxy-5-(trifluormethoxy)benzaldehyd 97%
2-Methoxy-5-(trifluoromethoxy)benzaldehyde
Formel:
C₉H₇F₃O₃ Dichte: 1,36 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115958 CAS-Nummer: 145742-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...