4-Methyl-2-(trifluoromethyl)benzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC10463-1G
PC10463-250MG
APOSPC10463-1GEA
719
EUR
APOSPC10463-1G
APOSPC10463-250MG
4-Methyl-2-(trifluoromethyl)benzaldehyde
4-Methyl-2-(trifluoromethyl)benzaldehyde
Formel:
C₉H₇F₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD18399857 CAS-Nummer: 1261622-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...