3,5-Difluor-2-pyridincarbonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Cyan-3,5-difluorpyridin, 3,5-Difluor-2-pyridincarbonsäurenitril
PC10521-25G
APOSPC10521-25GEA
75.4
EUR
APOSPC10521-25G
3,5-Difluor-2-pyridincarbonitril
3,5-Difluor-2-pyridincarbonitril
Formel:
C₆H₂F₂N₂ Molecular Weight: 140,09 g/mol Schmelzpunkt: 30…34 °C Flammpunkt: 90 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03788759 CAS-Nummer: 298709-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...