4-(Trifluoromethyl)phenylglyoxal hydrate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde hydrate, Oxo[4-(trifluoromethyl)phenyl]acetaldehyde hydrate (1:1)

PC10550-25G
APOSPC10550-25GEA 292 EUR
APOSPC10550-25G
4-(Trifluoromethyl)phenylglyoxal hydrate 97%
4-(Trifluoromethyl)phenylglyoxal hydrate
Formel: C₉H₅F₃O₂·xH₂O
Molecular Weight: 202.13 g/mol (anhy)
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664098
CAS-Nummer: 1736-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR