(4-Aminophenyl)[4-(2-fluorophenyl)piperazino]methanone

Lieferant: Apollo Scientific
Achtung

PC10557-1G
APOSPC10557-1GEA 186 EUR
APOSPC10557-1G
(4-Aminophenyl)[4-(2-fluorophenyl)piperazino]methanone
(4-Aminophenyl)[4-(2-fluorophenyl)piperazino]methanone
Formel: C₁₇H₁₈FN₃O
Molecular Weight: 299,35 g/mol
Schmelzpunkt: 161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659557
CAS-Nummer: 885949-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR