Pentafluorphenyl-2,4-dichlorbenzolsulfonat

Lieferant: Apollo Scientific

PC10569-1G PC10569-5G
APOSPC10569-1GEA 183 EUR
APOSPC10569-1G APOSPC10569-5G
Pentafluorphenyl-2,4-dichlorbenzolsulfonat
Pentafluorphenyl-2,4-dichlorbenzolsulfonat
Formel: C₁₂H₃Cl₂F₅O₃S
Schmelzpunkt: 79…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659543
CAS-Nummer: 885949-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR