Pentafluorphenyl-1-methansulfonat
Lieferant: Apollo Scientific
|
Achtung
|
PC10572-1G
APOSPC10572-1GEA
136
EUR
APOSPC10572-1G
Pentafluorphenyl-1-methansulfonat
Pentafluorphenyl-1-methansulfonat
Formel:
C₇H₃O₃F₅S Siedepunkt: 76 °C/0,6mmHg Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06659544 CAS-Nummer: 161912-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...