1-(2,3-Difluorphenyl)-2-thioharnstoff 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Difluorphenylthioharnstoff, N-(2,3-Difluorphenyl)thioharnstoff

PC10573-1G PC10573-5G PC10573-10G
APOSPC10573-1GEA 71.2 EUR
APOSPC10573-1G APOSPC10573-5G APOSPC10573-10G
1-(2,3-Difluorphenyl)-2-thioharnstoff 97%
1-(2,3-Difluorphenyl)-2-thioharnstoff
Formel: (C₆H₃F₂)NC(S)NH₂
Schmelzpunkt: 137…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368705
CAS-Nummer: 572889-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR