1-(2,6-Difluorphenyl)-2-thioharnstoff

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Difluorphenylthioharnstoff, N-(2,6-Difluorphenyl)thioharnstoff

PC10574-5G
APOSPC10574-5GEA 163 EUR
APOSPC10574-5G
1-(2,6-Difluorphenyl)-2-thioharnstoff
1-(2,6-Difluorphenyl)-2-thioharnstoff
Formel: (C₆H₃F₂)NC(S)NH₂
Molecular Weight: 188,2 g/mol
Schmelzpunkt: 151…153,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05022144
CAS-Nummer: 59772-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR