2-[(4-Fluorophenyl)sulphanyl]-5-nitrobenzenecarbaldehyde 95+%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluorophenylthio)-5-nitrobenzaldehyde

PC10577-1G
APOSPC10577-1GEA 154 EUR
APOSPC10577-1G
2-[(4-Fluorophenyl)sulphanyl]-5-nitrobenzenecarbaldehyde 95+%
2-[(4-Fluorophenyl)sulphanyl]-5-nitrobenzenecarbaldehyde
Formel: C₁₃H₈FNO₃S
Molecular Weight: 277,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07021301
CAS-Nummer: 35985-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR