2-[(4-Aminophenyl)sulphanyl]-N-(2,5-difluorophenyl)acetamide

Lieferant: Apollo Scientific

PC10580-1G
APOSPC10580-1GEA 93.1 EUR
APOSPC10580-1G
2-[(4-Aminophenyl)sulphanyl]-N-(2,5-difluorophenyl)acetamide
2-[(4-Aminophenyl)sulphanyl]-N-(2,5-difluorophenyl)acetamide
Formel: C₁₄H₁₂F₂N₂OS
Molecular Weight: 294,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07021305
CAS-Nummer: 885267-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR