4-(4-Fluorophenyl)-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulphide

Lieferant: Apollo Scientific

Synonyme: 4-(4-Fluorophenyl)-5-phenyl-4H-1,2,4-triazole-3-thiol

PC10585-1G
APOSPC10585-1GEA 93.1 EUR
APOSPC10585-1G
4-(4-Fluorophenyl)-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulphide
4-(4-Fluorophenyl)-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulphide
Formel: C₁₄H₁₀FN₃S
Molecular Weight: 271,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02720697
CAS-Nummer: 120873-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR