5-(3-Chlorophenyl)-4-(4-fluorophenyl)-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific

PC10587-1G
APOSPC10587-1GEA 32.1 EUR
APOSPC10587-1G
5-(3-Chlorophenyl)-4-(4-fluorophenyl)-4H-1,2,4-triazole-3-thiol
5-(3-Chlorophenyl)-4-(4-fluorophenyl)-4H-1,2,4-triazole-3-thiol
Formel: C₁₄H₉ClFN₃S
Molecular Weight: 305,758 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05150470
CAS-Nummer: 720667-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR