2-Amino-4,6-difluorobenzotrifluoride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluoro-2-(trifluoromethyl)aniline

PC10593-1G
APOSPC10593-1GEA 201 EUR
APOSPC10593-1G
2-Amino-4,6-difluorobenzotrifluoride 97%
2-Amino-4,6-difluorobenzotrifluoride
Formel: C₇H₄F₅N
Molecular Weight: 197,105 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972831
CAS-Nummer: 123973-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR