2-Iodo-5-(trifluoromethoxy)toluene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Iodo-2-methyl-4-(trifluoromethoxy)benzene, 4-Iodo-3-methyl-α,α,α-trifluoroanisole, 4-Iodo-3-methylphenyl trifluoromethyl ether
PC1061-5G
APOSPC1061-5GEA
172
EUR
APOSPC1061-5G
2-Iodo-5-(trifluoromethoxy)toluene
2-Iodo-5-(trifluoromethoxy)toluene
Formel:
C₈H₆F₃IO Molecular Weight: 302,032 g/mol Dichte: 1,776 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07368714 CAS-Nummer: 886763-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...