1,1,2,2-Tetrafluoro-3-methoxypropane

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2,2,3,3-tetrafluoropropyl ether

PC10613-25G
APOSPC10613-25GEA 352 EUR
APOSPC10613-25G
1,1,2,2-Tetrafluoro-3-methoxypropane
1,1,2,2-Tetrafluoro-3-methoxypropane
Formel: C₄H₆F₄O
Molecular Weight: 146,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075284
CAS-Nummer: 60598-17-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR