2-Chlor-3-nitro-5-(trifluormethyl)pyridin 98%

Lieferant: Apollo Scientific
Achtung

PC10641-25G
APOSPC10641-25GEA 163 EUR
APOSPC10641-25G
2-Chlor-3-nitro-5-(trifluormethyl)pyridin 98%
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD06255171
CAS-Nummer: 72587-15-6
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR