4-Chloro-5-fluoro-2-nitrophenol 98%

Lieferant: Apollo Scientific
Achtung

PC10643-1G PC10643-5G
APOSPC10643-1GEA 52.2 EUR
APOSPC10643-1G APOSPC10643-5G
4-Chloro-5-fluoro-2-nitrophenol 98%
4-Chloro-5-fluoro-2-nitrophenol
Formel: C₆H₃ClFNO₃
Lagertemperatur: Kühlschrank
MDL: MFCD11840294
CAS-Nummer: 345-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR