3-Bromo-5-fluoro-2-pyridine carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-2-carboxy-5-fluoropyridine, 3-Bromo-5-fluoropicolinic acid, 3-Bromo-5-fluoropyridine-2-carboxylic acid

PC10646-250MG
APOSPC10646-250MGEA 21.9 EUR
APOSPC10646-250MG
3-Bromo-5-fluoro-2-pyridine carboxylic acid 97%
3-Bromo-5-fluoro-2-pyridine carboxylic acid
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
MDL: MFCD12827684
CAS-Nummer: 1189513-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR