2-(Aminomethyl)-3-bromo-5-fluoropyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-5-fluoropyridin-2-yl)methylamine hydrochloride, 3-Bromo-5-fluoropicolinylamine hydrochloride

PC10647-100MG
APOSPC10647-100MGEA 154 EUR
APOSPC10647-100MG
2-(Aminomethyl)-3-bromo-5-fluoropyridine hydrochloride
2-(Aminomethyl)-3-bromo-5-fluoropyridine hydrochloride
Formel: C₆H₆BrFN₂·ClH
Molecular Weight: 241,489 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17214199
CAS-Nummer: 1257535-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR