6-Chloro-5-fluoronicotinonitrile 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-5-fluoropyridine-3-carbonitrile, 6-Chloro-3-cyano-5-fluoropyridine

PC10649-1G
APOSPC10649-1GEA 114 EUR
APOSPC10649-1G
6-Chloro-5-fluoronicotinonitrile 96%
6-Chloro-5-fluoronicotinonitrile
Formel: C₆H₂ClFN₂
Lagertemperatur: Kühlschrank
MDL: MFCD09972184
CAS-Nummer: 1020253-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR