3-Chlor-5-(trifluormethyl)picolinsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid, 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 3-Chloro-5-(trifluoromethyl)picolinic acid
PC10653-1G
APOSPC10653-1GEA
21.9
EUR
APOSPC10653-1G
3-Chlor-5-(trifluormethyl)picolinsäure 98%
3-Chlor-5-(trifluormethyl)picolinsäure
Formel:
C₇H₃ClF₃NO₂ Molecular Weight: 225,60 g/mol Schmelzpunkt: 135…138 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00277482 CAS-Nummer: 80194-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...