6-Fluoro-4-methylnicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Carboxy-2-fluoro-4-methylpyridine, 6-Fluoro-4-methylpyridine-3-carboxylic acid, 2-Fluoro-4-methylpyridine-5-carboxylic acid

PC10655-250MG
APOSPC10655-250MGEA 21.9 EUR
APOSPC10655-250MG
6-Fluoro-4-methylnicotinic acid
6-Fluoro-4-methylnicotinic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260452
CAS-Nummer: 944582-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR