3-Fluoro-6-methylpyridine-2-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Cyano-3-fluoro-6-methylpyridine, 3-Fluoro-6-methylpicolinonitrile

PC10657-500MG
APOSPC10657-500MGEA 321 EUR
APOSPC10657-500MG
3-Fluoro-6-methylpyridine-2-carbonitrile
3-Fluoro-6-methylpyridine-2-carbonitrile
Formel: C₇H₅FN₂
Molecular Weight: 136,126 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08062849
CAS-Nummer: 1211527-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR