2-(Aminomethyl)-3-fluoro-6-methylpyridine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Fluoro-6-methylpyridin-2-yl)methylamine hydrochloride, (3-Fluoro-6-methylpyridin-2-yl)methanamine hydrochloride
PC10659-1G
PC10659-250MG
APOSPC10659-1GEA
765
EUR
APOSPC10659-1G
APOSPC10659-250MG
2-(Aminomethyl)-3-fluoro-6-methylpyridine hydrochloride
2-(Aminomethyl)-3-fluoro-6-methylpyridine hydrochloride
Formel:
C₇H₉FN₂·ClH Lagertemperatur: Kühlschrank |
MDL:
MFCD17214195 CAS-Nummer: 1257535-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...