2-Brom-5-(trifluormethoxy)benzylbromid 95%

Lieferant: Apollo Scientific
Gefahr

PC1067-1G PC1067-5G
APOSPC1067-1GEA 132 EUR
APOSPC1067-1G APOSPC1067-5G
2-Brom-5-(trifluormethoxy)benzylbromid 95%
2-Brom-5-(trifluormethoxy)benzylbromid
Formel: C₈H₅Br₂F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD07368721
CAS-Nummer: 886763-13-9
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR