3-Bromo-4-(trifluoromethyl)benzaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-formylbenzotrifluoride, 3-Bromo-α,α,α-trifluoro-p-tolualdehyde
PC10671-10G
PC10671-1G
PC10671-5G
PC10671-25G
APOSPC10671-10GEA
541
EUR
APOSPC10671-10G
APOSPC10671-1G
APOSPC10671-5G
APOSPC10671-25G
3-Bromo-4-(trifluoromethyl)benzaldehyde 98%
3-Bromo-4-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄BrF₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185384 CAS-Nummer: 372120-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...