2-Iodo-4-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC10681-1G PC10681-250MG PC10681-5G
APOSPC10681-1GEA 0 EUR
APOSPC10681-1G APOSPC10681-250MG APOSPC10681-5G
2-Iodo-4-(trifluoromethyl)benzyl bromide 97%
2-Iodo-4-(trifluoromethyl)benzyl bromide
Formel: C₈H₅BrF₃I
Lagertemperatur: Raumtemperatur
MDL: MFCD08543940
CAS-Nummer: 939758-31-3
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR