1-Fluoro-3,3-dimethyl-1,2-benziodoxole 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC10688-1G
APOSPC10688-1GEA
143
EUR
APOSPC10688-1G
1-Fluoro-3,3-dimethyl-1,2-benziodoxole 98%
1-Fluoro-3,3-dimethyl-1,2-benziodoxole
Formel:
C₉H₁₀FIO Molecular Weight: 280,08 g/mol Schmelzpunkt: 88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28397076 CAS-Nummer: 1391728-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...