2-Bromo-5-(trifluoromethoxy)benzyl alcohol 95%

Lieferant: Apollo Scientific

PC1069-1G PC1069-5G
APOSPC1069-1GEA 0 EUR
APOSPC1069-1G APOSPC1069-5G
2-Bromo-5-(trifluoromethoxy)benzyl alcohol 95%
2-Bromo-5-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07368723
CAS-Nummer: 886763-15-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR