N-Methyl-N-(trifluoromethylthio)aniline 98%

Lieferant: Apollo Scientific

PC10690-1G
APOSPC10690-1GEA 469 EUR
APOSPC10690-1G
N-Methyl-N-(trifluoromethylthio)aniline 98%
N-Methyl-N-(trifluoromethylthio)aniline
Formel: C₈H₈F₃NS
Molecular Weight: 207,21 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 66476-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR